MilliporeSigma
  • Metabolomic analysis of avocado fruits by GC-APCI-TOF MS: effects of ripening degrees and fruit varieties.
Metabolomic analysis of avocado fruits by GC-APCI-TOF MS: effects of ripening degrees and fruit varieties.
Analytical and bioanalytical chemistry (2014-11-10)
E Hurtado-Fernández, T Pacchiarotta, O A Mayboroda, A Fernández-Gutiérrez, A Carrasco-Pancorbo
ABSTRACT
In order to investigate avocado fruit ripening, nontargeted GC-APCI-TOF MS metabolic profiling analyses were carried out. Principal component analysis (PCA) and partial least squares discriminant analysis (PLS-DA) were used to explore the metabolic profiles from fruit samples of 13 varieties at two different ripening degrees. Mannoheptulose; pentadecylfuran; aspartic, malic, stearic, citric and pantothenic acids; mannitol; and β-sitosterol were some of the metabolites found as more influential for the PLS-DA model. The similarities among genetically related samples (putative mutants of "Hass") and their metabolic differences from the rest of the varieties under study have also been evaluated. The achieved results reveal new insights into avocado fruit composition and metabolite changes, demonstrating therefore the value of metabolomics as a functional genomics tool in characterizing the mechanism of fruit ripening development, a key developmental stage in most economically important fruit crops.
Product Description
Supelco
Metolachlor OA, PESTANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyridine, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Pyridine, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide, for GC derivatization, LiChropur, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
SKU
Pack Size
Availability
Price
Quantity
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)