Merck
  • Quantification of nitropropanoyl glucosides in karaka nuts before and after treatment.
Quantification of nitropropanoyl glucosides in karaka nuts before and after treatment.
Food chemistry (2015-01-13)
J J MacAskill, M Manley-Harris, Richard J Field
ABSTRACT
A high performance liquid chromatography (HPLC) method was developed to assay nitropropanoyl glucosides in the nuts of karaka (Corynocarpus laevigatus) a traditional food of New Zealand Māori. Levels of glucosides, measured as 3-nitropropanoic acid, ranged from 50.25 to 138.62 g kg(-1) (5.0-13.9% w/w) and were highest in nuts from unripe drupes; these levels are higher than any previously reported. Other parts of the drupe also contained nitropropanoyl glucosides but at lower levels than the nut. Treatment procedures to remove the glucosides from the nuts varied in their efficacy with soxhlet extraction removing 98.7% and prolonged boiling and cold water extraction both removing 96%. These findings confirm the traditional methods for preparation of these nuts for consumption.
Product Description
Supelco
Malonic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
3-Nitropropionic acid, 97%
Sigma-Aldrich
3-Nitropropionic acid, ≥97%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Malonic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC