Merck
  • Vigabatrin in dried plasma spots: validation of a novel LC-MS/MS method and application to clinical practice.
Vigabatrin in dried plasma spots: validation of a novel LC-MS/MS method and application to clinical practice.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-06-09)
Nađa Kostić, Yannis Dotsikas, Nebojša Jović, Galina Stevanović, Anđelija Malenović, Mirjana Medenica
ABSTRACT
This paper presents a LC-MS/MS method for the determination of antiepileptic drug vigabatrin in dried plasma spots (DPS). Due to its zwitterionic chemical structure, a pre-column derivatization procedure was performed, aiming to yield enhanced ionization efficiency and improved chromatographic behaviour. Propyl chloroformate, in the presence of propanol, was selected as the best derivatization reagent, providing a strong signal along with reasonable run time. A relatively novel sample collection technique, DPS, was utilized, offering easy sample handling and analysis, using a sample in micro amount (∼5μL). Derivatized vigabatrin and its internal standard, 4-aminocyclohexanecarboxylic acid, were extracted by liquid-liquid extraction (LLE) and determined in positive ion mode by applying two SRM transitions per analyte. A Zorbax Eclipse XDB-C8 column (150×4.6mm, 5μm particle size) maintained at 30°C, was utilized with running mobile phase composed of acetonitrile: 0.15% formic acid (85:15, v/v). Flow rate was 550μL/min and total run time 4.5min. The assay exhibited excellent linearity over the concentration range of 0.500-50.0μg/mL, which is suitable for the determination of vigabatrin level after per os administration in children and youths with epilepsy, who were on vigabatrin therapy, with or without co-medication. Specificity, accuracy, precision, recovery, matrix-effect and stability were also estimated and assessed within acceptance criteria.
Product Description
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
4-Aminocyclohexanecarboxylic acid, AldrichCPR
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%