Merck
  • Comparison of a novel TiO₂/diatomite composite and pure TiO₂ for the purification of phosvitin phosphopeptides.
Comparison of a novel TiO₂/diatomite composite and pure TiO₂ for the purification of phosvitin phosphopeptides.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-05-03)
Yang Zhang, Junhua Li, Fuge Niu, Jun Sun, Yuan Dou, Yuntao Liu, Yujie Su, Bei Zhou, Qinqin Xu, Yanjun Yang
ABSTRACT
A novel TiO2/diatomite composite (TD) was prepared and then characterized by scanning electron microscope (SEM) and Fourier Transform Infrared (FTIR). The results of SEM showed that after modification, the porous surface of diatomite was covered with TiO2. Both diatomite and TD had clear disc-shaped structures with average grain diameters of around 25 μm. Then TD and pure TiO2 were applied in the purification of phosvitin phosphopeptides (PPPs) from the digest of egg yolk protein, and a comparative study of adsorption properties of PPPs on TD and TiO2 was performed. In the study of adsorption kinetics, the adsorption equilibrium of PPPs on TD and TiO2 fitted well with the Langmuir model, and the time needed to reach adsorption equilibrium were both around 10 min. The maximum dynamic adsorption capacity of TD (8.15 mg/g) was higher than that of TiO2 (4.96 mg/g). The results of repeated use showed that TD and TiO2 were very stable after being subjected to ten repeated adsorption-elution cycles, and TD could easily be separated from aqueous solution by filtration. On the other hand, the present synthetic technology of TD was very simple, cost-effective, organic solvent-free and available for large-scale preparation. Thus, this separation method not only brings great advantages in the purification of PPPs from egg yolk protein but also provides a promising purification material for the enrichment of phosphopeptides in proteomic researches.
Product Description
Supelco
Acetone, analytical standard
Sigma-Aldrich
Copper(II) sulfate pentahydrate, purum p.a., crystallized, ≥99.0% (RT)
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium sulfite, BioXtra, ≥98%
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammonium molybdate, meets USP testing specifications
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Sodium sulfite, BioUltra, anhydrous, ≥98% (RT)