Merck
  • Expression kinetics of metalloproteinases and their tissue inhibitors in experimental murine pulmonary tuberculosis.
Expression kinetics of metalloproteinases and their tissue inhibitors in experimental murine pulmonary tuberculosis.
Experimental lung research (2014-10-03)
Ana G Ramos-Martínez, José A Enciso-Moreno, Israel Espinosa-Ayala, Dulce Mata-Espinoza, Bruno Rivas-Santiago, Valentín Trujillo-Paez, Joel Monárrez-Espino, Rogelio Hernández-Pando, Carmen J Serrano
ABSTRACT
Explore the temporal expression of metalloproteinases (MMPs) and tissue inhibitors of metalloproteinases (TIMPs) during experimental tuberculosis induced by virulent Mycobacterium tuberculosis strain H37Rv. BALB/c mice were infected via endotracheal instillation with H37Rv. Groups of mice were euthanized at different time points during infection. RNA was isolated from the lungs, and the expression of MMP-3, 8, 9, 10, 12, 13 and TIMP-1-4 was determined by quantitative PCR. Immunohistochemical detection of MMP-3, MMP-9, and MMP-10 was done to determine the cell source. The infection with H37Rv-induced inflammation resulted in maximal up-regulation of MMP-3, 8, 9, 10, 12 and 13 at day 21 postinfection. Additionally, MMP-13 showed another expression peak during late disease at day 60. Airway epithelium and macrophages were the most common MMP-3 and MMP-9 immunopositive cells, while for MMP-10, macrophages and endothelial cells were the most common, particularly at days 14 and 21 in well-formed granulomas. During late disease, vacuolated macrophages in pneumonic areas and bronchial epithelium showed mild MMP immunostaining. MMP-3, 8, 9, 10, 12, and 13 are maximally expressed at the peak of granuloma formation in the mouse tuberculosis model, with no compensation in levels or timing of TIMP expression. This data opens the possibility of participation of these molecules in the granuloma process.
Product Description
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Supelco
Oleic acid, Selectophore, ≥99%
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, FCC, FG