Merck
  • Flexible membranes of MoS2/C nanofibers by electrospinning as binder-free anodes for high-performance sodium-ion batteries.
Flexible membranes of MoS2/C nanofibers by electrospinning as binder-free anodes for high-performance sodium-ion batteries.
Scientific reports (2015-03-26)
Xiaoqin Xiong, Wei Luo, Xianluo Hu, Chaoji Chen, Long Qie, Dongfang Hou, Yunhui Huang
ABSTRACT
A flexible membrane consisting of MoS2/carbon nanofibers has been fabricated by a simple electrospinning method. MoS2 nanosheets are uniformly encapsulated in the inter-connected carbon nanofibers with diameters of ~150 nm. When evaluated as a binder-free electrode for sodium-ion batteries, the as-obtained electrode demonstrates high performances, including high reversible capacity of 381.7 mA h g(-1) at 100 mA g(-1) and superior rate capability (283.3, 246.5 and 186.3 mA h g(-1) at 0.5, 1 and 2 A g(-1), respectively). Most importantly, the binder-free electrode made of MoS2 and carbon nanofibers can still deliver a charge capacity of 283.9 mA h g(-1) after 600 cycles at a current density of 100 m A g(-1), indicating a very promising anode for long-life SIBs.
Sigma-Aldrich
Ammonium tetrathiomolybdate, 99.97% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%