Merck
  • Effect of phosphomimetic mutations on the C-terminal sensitivity of glycine transporter GlyT1 to calpain.
Effect of phosphomimetic mutations on the C-terminal sensitivity of glycine transporter GlyT1 to calpain.
Neuroscience research (2014-02-26)
Andrea Mihalikova, Martina Baliova, Frantisek Jursky
ABSTRACT
Previously, we found that the C-terminus of the glycine transporter GlyT1 loses the most of its epitopes during pathological calcium increase in rodent synaptosomes but that the more internal epitopes are spared. We also found that epitope immunoreactivity likely decreases via both phosphorylation and calpain-mediated proteolysis. Here we show that the predicted phosphomimetic mutation S605D fully blocks the adjacent (T602/T603) internal calpain cleavage in the mouse GlyT1b C-terminal fusion protein, but that the neutral S605A mutation does not. Consistent with this, the phophomimetic mutation, but not the neutral mutation, significantly protected the internal GlyT1 C-terminal antiGlyT1C603-626 epitopes against calpain when introduced into tissue culture expressed GlyT1b. Because similar effects can be obtained using phosphatase inhibitors, it may be that phosphorylation of S605 protects the GlyT1 C-terminal sequences from calpain cleavage in vivo.
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Alanine, European Pharmacopoeia (EP) Reference Standard
SAFC
HEPES
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
SAFC
HEPES
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Trometamol, European Pharmacopoeia (EP) Reference Standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane