Merck
  • Exploring the interactions of decabrominateddiphenyl ether and tetrabromobisphenol A with human serum albumin.
Exploring the interactions of decabrominateddiphenyl ether and tetrabromobisphenol A with human serum albumin.
Environmental toxicology and pharmacology (2014-09-10)
Yan-Qing Wang, Hong-Mei Zhang, Jian Cao
ABSTRACT
Decabrominateddiphenyl ether (deca-BDE) and tetrabromobisphenol A (TBBPA) are known as brominated flame-retardants, which are commonly found in the environment. The binding mechanisms of deca-BDE and TBBPA with human serum albumin (HSA) are still unknown. In this report, the interactions of deca-BDE and TBBPA with HSA were investigated using different spectroscopic methods and molecular modeling. The experimental results indicated the formation of complexes between deca-BDE/TBBPA and HSA with different affinity. These interactions affected the secondary structure of HSA. Thermodynamic investigations revealed that hydrophobic forces mainly drove the binding interactions of deca-BDE/TBBPA with HSA. For TBBPA, hydrogen-bonding interactions were also involved in the binding process of TBBPA with HSA. According to the analysis of experimental and theoretical data, we concluded that the binding site of deca-BDE to HSA located in the subdomain IB, while TBBPA was near to subdomain IIA and Trp-214. The binding interactions of deca-BDE and TBBPA with the most prominent carrier protein in the human circulatory system could influence mechanisms of their biochemical processes. Thus, these binding interactions can play central roles in studying the distribution and toxicity mechanisms of brominated flame-retardants.
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
3,3′,5,5′-Tetrabromobisphenol A, 97%
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Supelco
Ibuprofen