Merck
  • Salt stability--effect of particle size, relative humidity, temperature and composition on salt to free base conversion.
Salt stability--effect of particle size, relative humidity, temperature and composition on salt to free base conversion.
Pharmaceutical research (2014-08-26)
Yi-Ling Hsieh, Lynne S Taylor
ABSTRACT
The aim of this study was to investigate how factors such as temperature, relative humidity and particle size impact the extent of disproportionation (salt to free base conversion) in powder blends of miconazole, benzocaine or sertraline mesylate salts mixed with a basic additive. Raman spectroscopy was used to quantitate the extent of disproportionation. The data was further analyzed by multivariate analysis with partial least squares (PLS) modeling. It was found that salt disproportionation was significantly influenced by % weight gain due to moisture sorption both in terms of the kinetics and the conversion extent, suggesting a solution-mediated reaction. Temperature plays an important role in impacting the value of pHmax which in turn has a significant correlation to the amount of free base formed. The particle size and drug: additive ratio were also found to influence the extent of disproportionation. This study shows that the extent of salt disproportionation is influenced by multiple factors and the application of PLS modeling demonstrated the feasibility of utilizing multivariate analysis to generate a predictive model for estimating the extent of conversion and thus may serve as a tool for risk assessment.
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Magnesium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium chloride hexahydrate, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride hexahydrate, tested according to Ph. Eur.
USP
Miconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Magnesium chloride hexahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E511, 99-101%, ≤0.0001% Al
Miconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride hexahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium nitrate hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Magnesium nitrate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet