Merck
  • Inhibition of influenza virus replication by targeting broad host cell pathways.
Inhibition of influenza virus replication by targeting broad host cell pathways.
PloS one (2014-10-22)
Isabelle Marois, Alexandre Cloutier, Isabelle Meunier, Hana M Weingartl, André M Cantin, Martin V Richter
ABSTRACT
Antivirals that are currently used to treat influenza virus infections target components of the virus which can mutate rapidly. Consequently, there has been an increase in the number of resistant strains to one or many antivirals in recent years. Here we compared the antiviral effects of lysosomotropic alkalinizing agents (LAAs) and calcium modulators (CMs), which interfere with crucial events in the influenza virus replication cycle, against avian, swine, and human viruses of different subtypes in MDCK cells. We observed that treatment with LAAs, CMs, or a combination of both, significantly inhibited viral replication. Moreover, the drugs were effective even when they were administered 8 h after infection. Finally, analysis of the expression of viral acidic polymerase (PA) revealed that both drugs classes interfered with early events in the viral replication cycle. This study demonstrates that targeting broad host cellular pathways can be an efficient strategy to inhibit influenza replication. Furthermore, it provides an interesting avenue for drug development where resistance by the virus might be reduced since the virus is not targeted directly.
Sigma-Aldrich
Mefloquine hydrochloride, ≥98% (HPLC), powder
Supelco
Capsaicin, analytical standard
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Sigma-Aldrich
Primaquine bisphosphate, 98%
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
Quinacrine dihydrochloride, ≥90%
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Calcium Ionophore A23187 mixed calcium magnesium salt, Approximate 1:15 molar ratio Ca:Mg.
Quinine sulfate, meets USP testing specifications, monograph mol wt. 782.94 ((C20H24N2O2)2 . H2SO4 . 2H2O)
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Primaquine diphosphate, European Pharmacopoeia (EP) Reference Standard
Mefloquine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Capsaicin, United States Pharmacopeia (USP) Reference Standard
Supelco
Capsaicin, Pharmaceutical Secondary Standard; Certified Reference Material
Capsaicin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Quinine hemisulfate salt monohydrate, tested according to Ph. Eur.
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Sigma-Aldrich
Quinine hemisulfate salt monohydrate, BioReagent, suitable for fluorescence, 99.0-101.0%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Quinine hemisulfate salt monohydrate, synthetic, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Amodiaquin dihydrochloride dihydrate, analytical standard