Merck
  • Direct doping analysis of beta-blocker drugs from urinary samples by on-line molecularly imprinted solid-phase extraction coupled to liquid chromatography/mass spectrometry.
Direct doping analysis of beta-blocker drugs from urinary samples by on-line molecularly imprinted solid-phase extraction coupled to liquid chromatography/mass spectrometry.
The Analyst (2015-02-14)
Mariane Gonçalves Santos, Isabela Maria Campos Tavares, Vanessa Bergamin Boralli, Eduardo Costa Figueiredo
ABSTRACT
The use of beta-blockers to enhance performance in some sports is forbidden. Based on this regulation, there is a demand for dynamic analytical procedures for analyzing these compounds quickly and without manual sample preparation. Therefore, the use of a molecularly imprinted polymer (MIP) in a multidimensional liquid chromatographic system coupled to a mass spectrometer provides a good alternative for improving the selectivity and practicality of the beta-blocker analyses, as described in this paper. A water-compatible MIP for oxprenolol was synthesized by the precipitation method, using methacrylic acid as a functional monomer and 2-hydroxyethyl methacrylate and glycerol dimethacrylate as hydrophilic monomers. A column filled with MIP was coupled to an LC-MS/MS instrument under the multidimensional configuration, with 10.0 mmol L(-1) ammonium formate buffer (pH 5.0) as the loading and reconditioning mobile phase and a 0.01% formic acid aqueous solution-methanol (30 : 70 v : v) as the elution mobile phase. The system was used for on-line extraction and quantization of oxprenolol (from 1.0 to 75.0 μg L(-1)), atenolol, propranolol, nadolol, pindolol, labetalol and metoprolol (all from 3.0 to 50 μg L(-1)) simultaneously, from urine samples. The correlation coefficient was higher than 0.99 for all the analytes. Suitable precision and accuracy were obtained.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Atenolol, European Pharmacopoeia (EP) Reference Standard
Pindolol, European Pharmacopoeia (EP) Reference Standard
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Atenolol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%