Merck
  • Effects of CO2 plant extracts on triacylglycerol oxidation in Atlantic salmon during cooking and storage.
Effects of CO2 plant extracts on triacylglycerol oxidation in Atlantic salmon during cooking and storage.
Food chemistry (2014-12-04)
Marko Tarvainen, Anu Nuora, Karl-Werner Quirin, Heikki Kallio, Baoru Yang
ABSTRACT
Increasing concern of consumers on the safety of synthetic food additives has created high interest in natural preservatives in food industry. Plant extracts produced by supercritical CO2 technology from rosemary (R), oregano (O) and an antimicrobial blend (AB) consisting of seven different plants were studied for their effects on lipid oxidation in Atlantic salmon (Salmo salar). Fish pieces were marinated with rapeseed oil containing 0, 1, 2 or 4 g of plant extracts/kg of fish. After cooking the pieces were stored in refrigerator for 26 days. Peroxide values (PVs) were determined and oxidised triacylglycerols (TAGs) measured by UHPLC-ESI/MS at 0, 7, 14 and 26 days of storage. During the first two weeks of storage, AB delayed oxidation by at least one week compared to control samples as shown by PVs (<10 meq. O2) and by the oxidised TAGs. Oregano and rosemary showed also some antioxidative potential.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Tripentadecanoin, ≥99%
Supelco
Acetone, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%