Merck
  • Study of the catabolism of thyme phenols combining in vitro fermentation and human intervention.
Study of the catabolism of thyme phenols combining in vitro fermentation and human intervention.
Journal of agricultural and food chemistry (2014-10-24)
Juana I Mosele, Sandra Martín-Peláez, Alba Macià, Marta Farràs, Rosa-Maria Valls, Úrsula Catalán, María-José Motilva
ABSTRACT
The gut metabolism of four thyme phenolics (monoterpenes thymol and carvacrol, rosmarinic acid, and eriodictyol) was evaluated in vitro. After the in vitro transformations of the individual phenols had been studied, the presence of their microbial metabolites was investigated in human feces collected before and after a sustained intake (3 weeks) of 25 mL/day of a thyme phenol-enriched olive oil. Results of in vitro fermentation showed low degradation of thymol and carvacrol. By contrast, large catabolism was noted when rosmarinic acid and eriodictyol were fermented, yielding hydroxyphenylpropionic acid as the main metabolite. In accordance with these results, after the in vivo intervention with thyme phenol-enriched olive oil, an increase in the concentration of hydroxyphenylpropionic and phenylpropionic acids was observed in human feces, confirming the effective in vivo microbial degradation of rosmarinic acid and eriodictyol. Carvacrol was detected in fecal samples at trace levels, suggesting that monoterpenes are well absorbed in the upper part of the gastrointestinal tract.
Sigma-Aldrich
Thymol, FCC, FG
Supelco
Thymol, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Thymol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Thymol, analytical standard
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Sigma-Aldrich
Thymol, ≥98.5%
Thymol, European Pharmacopoeia (EP) Reference Standard
Rosmarinic acid, primary reference standard
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Sigma-Aldrich
Rosmarinic acid, 96%
Rosmarinic acid, European Pharmacopoeia (EP) Reference Standard