Merck
  • Macrolide resistance conferred by rRNA mutations in field isolates of Mannheimia haemolytica and Pasteurella multocida.
Macrolide resistance conferred by rRNA mutations in field isolates of Mannheimia haemolytica and Pasteurella multocida.
The Journal of antimicrobial chemotherapy (2014-09-28)
Anders S Olsen, Ralf Warrass, Stephen Douthwaite
ABSTRACT
To determine how resistance to macrolides is conferred in field isolates of Pasteurella multocida and Mannheimia haemolytica that lack previously identified resistance determinants for rRNA methylation, efflux and macrolide-modifying enzymes. Isolates of P. multocida and M. haemolytica identified as being highly resistant (MICs >64 mg/L) to the macrolides erythromycin, gamithromycin, tilmicosin, tildipirosin and tulathromycin were screened by multiplex PCR for the previously identified resistance genes erm(42), msr(E) and mph(E). Strains lacking these determinants were analysed by genome sequencing and primer extension on the rRNAs. Macrolide resistance in one M. haemolytica isolate was conferred by the 23S rRNA mutation A2058G; resistance in three P. multocida isolates were caused by mutations at the neighbouring nucleotide A2059G. In each strain, all six copies of the rrn operons encoded the respective mutations. There were no mutations in the ribosomal protein genes rplD or rplV, and no other macrolide resistance mechanism was evident. High-level macrolide resistance can arise from 23S rRNA mutations in P. multocida and M. haemolytica despite their multiple copies of rrn. Selective pressures from exposure to different macrolide or lincosamide drugs presumably resulted in consolidation of either the A2058G or the A2059G mutation.
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard