Merck
  • Inflammasomes Induced by 7-Ketocholesterol and Other Stimuli in RPE and in Bone Marrow-Derived Cells Differ Markedly in Their Production of IL-1β and IL-18.
Inflammasomes Induced by 7-Ketocholesterol and Other Stimuli in RPE and in Bone Marrow-Derived Cells Differ Markedly in Their Production of IL-1β and IL-18.
Investigative ophthalmology & visual science (2015-02-14)
Guangpu Shi, Siqi Chen, Wambui S Wandu, Osato Ogbeifun, Lindsey F Nugent, Arvydas Maminishkis, Samuel J H Hinshaw, Ignacio R Rodriguez, Igal Gery
ABSTRACT
The inflammatory process plays a major role in the pathogenesis of AMD, and recent data indicate the involvement of inflammasomes. Inflammasomes are intracellular structures that trigger inflammation by producing mature interleukin-(IL)-1β and IL-18. This study examined the capacity of 7-ketocholesterol (7KCh), an oxysterol that accumulates in the retinal pigmented epithelium (RPE) and choroid, to initiate inflammasome formation in RPE and bone marrow-derived cells. Tested cells included fetal human RPE (fhRPE), human ARPE-19 cells, primary human brain microglia cells, and human THP-1 monocyte cells. 7-Ketocholesterol and other compounds were added to the cell cultures, and their stimulatory effects were determined by quantitative PCR and release of cytokines, measured by ELISA and Western blotting. 7-Ketocholesterol efficiently induced inflammasome formation by all primed cell populations, but secreted cytokine levels were higher in cultures of bone marrow-derived cells (microglia and THP-1 cells) than in RPE cultures. Interestingly, inflammasomes formed in cells of the two populations differed strikingly in their preferential production of the two cytokines. Thus, whereas bone marrow-derived cells produced levels of IL-1β that were higher than those of IL-18, the opposite was found with RPE cells, which secreted higher levels of IL-18. Importantly, Western blot analysis showed that IL-18, but not IL-1β, was expressed constitutively by RPE cells. 7-Ketocholesterol efficiently stimulates inflammasome formation and is conceivably involved in the pathogenesis of AMD. In contrast to bone marrow-derived cells, RPE cells produced higher levels of IL-18 than IL-1β. Further, IL-18, a multifunctional cytokine, was expressed constitutively by RPE cells. These observations provide new information about stimuli and cells and their products assumed to be involved in the pathogenesis of AMD.
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
5-Cholesten-3β-ol-7-one, ≥90%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Pricing and availability is not currently available. Contact Technical Service
SAFC
BIS-TRIS
Pricing and availability is not currently available. Contact Technical Service
SAFC
BIS-TRIS
Pricing and availability is not currently available. Contact Technical Service