Merck
  • mTOR inhibition improves fibroblast growth factor receptor targeting in hepatocellular carcinoma.
mTOR inhibition improves fibroblast growth factor receptor targeting in hepatocellular carcinoma.
British journal of cancer (2015-02-18)
T Scheller, C Hellerbrand, C Moser, K Schmidt, A Kroemer, S M Brunner, H J Schlitt, E K Geissler, S A Lang
ABSTRACT
Systemic therapy has proven only marginal effects in hepatocellular carcinoma (HCC) so far. The aim of this study was to evaluate the effect of targeting fibroblast growth factor receptor (FGFR) on tumour and stromal cells in HCC models. Human and murine HCC cells, endothelial cells (ECs), vascular smooth muscle cells (VSMCs), hepatic stellate cells (HSCs), human HCC samples, FGFR inhibitor BGJ398 and mammalian target of rapamycin (mTOR) inhibitor rapamycin were used. Effects on growth, motility, signalling and angiogenic markers were determined. In vivo subcutaneous and syngeneic orthotopic tumour models were used. In tumour cells and ECs, targeting FGFR showed significant inhibitory effects on signalling and motility. Minor effects of FGFR inhibition were observed on VSMCs and HSCs, which were significantly enhanced by combining FGFR and mTOR blockade. In vivo daily (5 mg kg(-1)) treatment with BGJ398 led to a significant growth inhibition in subcutaneous tumour models, but only a combination of FGFR and mTOR blockade impaired tumour growth in the orthotopic model. This was paralleled by reduced tumour cell proliferation, vascularisation, pericytes and increased apoptosis. Targeting FGFR with BGJ398 affects tumour cells and ECs, whereas only a combination with mTOR inhibition impairs recruitment of VSMCs and HSCs. Therefore, this study provides evidence for combined FGFR/mTOR inhibition in HCC.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Chloroform, analytical standard
Supelco
Acetone, analytical standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material