Merck
  • PET imaging of Oatp-mediated hepatobiliary transport of [(11)C] rosuvastatin in the rat.
PET imaging of Oatp-mediated hepatobiliary transport of [(11)C] rosuvastatin in the rat.
Molecular pharmaceutics (2014-06-25)
Jiake He, Yang Yu, Bhagwat Prasad, Jeanne Link, Robert S Miyaoka, Xijing Chen, Jashvant D Unadkat
ABSTRACT
A novel positron emission tomography (PET) tracer, [(11)C]-rosuvastatin (RSV), was developed to dynamically and noninvasively measure hepatobiliary transport and tissue distribution of [(11)C]-RSV in rats. Male Sprague-Dawley rats were administered either an Oatp inhibitor, rifampin (RIF, 40 mg/kg iv bolus plus 1.85 mg/min/kg iv infusion, n = 3), or the corresponding vehicle (saline, n = 3) for at least 90 min. Then, while these infusions were ongoing, the animals received [(11)C]-rosuvastatin (∼1 mCi/30 s, iv infusion). After [(11)C]-RSV administration, the lower abdominal region of the rats was imaged for 90 min. Time-activity curves for liver, intestine, and kidney were obtained and corrected for vascular content prior to noncompartmental and compartmental (five-compartment model) analysis. The majority of the [(11)C]-RSV dose was distributed into the liver. In the presence of RIF, the area under the [(11)C]-RSV radioactivity blood concentration-time profile (AUC0-90 min) was increased by ∼3-fold. Relative to the control animals, RIF reduced the distribution of [(11)C]-RSV radioactivity into the liver and kidney (tissue AUC0-15 min/blood AUC0-15 min) by 54% and 73% respectively. Compartmental modeling showed that RIF decreased CLBL, CLLI, CLBK, and CLK0 but had no effect on CLLB, where B, L, I, K, and 0 represent blood, liver, intestine, kidney, and irreversible loss. [(11)C]-RSV can be used to dynamically and noninvasively quantify hepatobiliary transport and hepatic concentration of the drug, in the absence and presence of drug interactions, in rats and could be used for the same purpose in humans.
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%