Merck
  • Preventive and therapeutic effects of a beta adrenoreceptor agonist, dobutamine, in carrageenan-induced inflammatory nociception in rats.
Preventive and therapeutic effects of a beta adrenoreceptor agonist, dobutamine, in carrageenan-induced inflammatory nociception in rats.
Inflammation (2014-05-09)
Tufan Mert, Berin Tugtag, Metin Kilinc, Elif Sahin, Hafize Oksuz, Yasemin Gunes
ABSTRACT
We hypothesized that locally administrated β-adrenoreceptor agonist can modulate the inflammatory nociceptive parameters in carrageenan (CG)-induced peripheral inflammatory pain. This study was therefore aimed to assess the preventive and therapeutic effects of a β-agonist, dobutamine, by investigating its pretreatment and posttreatment actions on the inflammation-induced hypersensitivities (thermal hyperalgesia, mechanical allodynia) to cutaneous stimulation, edema, and several biochemical oxidant and anti-oxidant parameters in a rat model of CG-induced hind paw inflammation. Effects of dobutamine were compared with those of esmolol, a β-adrenoreceptor antagonist. CG injection to healthy rats lowered the thermal latencies (from 10.1 ± 0.2 to 4.9 ± 0.1 s) and mechanical thresholds (from 32.9 ± 0.5 to 18.9 ± 1.3 g) and caused the hyperalgesia and allodynia. In CG-induced inflamed paws, while intraplantar esmolol treatment (1 mg) produced significant decreases in latencies (4.1 ± 0.1 s) and thresholds (15.2 ± 2.4 g), dobutamine (1 mg) increased the latencies (11.3 ± 0.5 s) and thresholds (26.3 ± 2.8 g). In contrast to esmolol, dobutamine increased the superoxide dismutase level and decreased the myeloperoxidase, malondialdehyde, and nitric oxide levels in CG-induced inflamed paws. The present results can reveal that β-adrenoreceptors may play a role in inflammatory nociceptive processes, and locally treated β-adrenoreceptor agonists such as dobutamine can be a preferable, appropriate choice for the management of inflammatory nociception due to their preventive and therapeutic effects on CG-induced peripheral inflammatory nociception.
Product Description
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Dobutamine hydrochloride, ≥98%
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Dobutamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Supelco
Sodium nitrite, analytical standard
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%