Merck
  • Lipopolysaccharide exposure during pregnancy leads to aortic dysfunction in offspring rats.
Lipopolysaccharide exposure during pregnancy leads to aortic dysfunction in offspring rats.
PloS one (2014-07-16)
Shanyu Zhao, Haigang Zhang, Dayan Cao, Ya Liu, Xiaohui Li
ABSTRACT
Prenatal exposure to Lipopolysaccharide (LPS) produces hypertension in adult offspring rats. The present study was to explore the effects of prenatal inflammation on morphological and functional changes in the aorta from offspring rats and to further assess its susceptibility to cardiovascular diseases. Pregnant rats were treated intraperitoneally on gestation Days 8, 10 and 12 with saline, LPS (0.79 mg/kg), or pyrrolidine dithiocarbamate (PDTC, 100 mg/kg)+LPS, respectively. Aortic ring reactivity and histopathological alteration were analyzed in offspring at the age of 12 weeks. The detections of connexin (Cx) 37, Cx40, Cx43, and Cx45, including immunofluorescent patterns, protein levels and mRNA expression in the aorta, were performed as well. Furthermore, the expressions of Nuclear factor (NF)-κB (p65), IκBα, phospho-IκBα and IκBβ were determined. The results showed that prenatal LPS exposure leads to morphological abnormalities and impaired aortic reactivity in offspring. Prenatal LPS exposure also decreased the protein and mRNA expression of Cx37 in the aorta from offspring rats. NF-κB and phospho-IκBα levels were both increased, IκBα level, however, was decreased in the aorta of offspring from the maternal LPS exposure compared to the controls. Simultaneously, PDTC treatment markedly reversed the action of LPS. Decreased expression of Cx37 contributed to the aortic dysfunction of prenatal LPS exposure offspring, which should be associated with NF-κB activation.
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%