Merck
  • Carbonic anhydrase activation is associated with worsened pathological remodeling in human ischemic diabetic cardiomyopathy.
Carbonic anhydrase activation is associated with worsened pathological remodeling in human ischemic diabetic cardiomyopathy.
Journal of the American Heart Association (2014-03-29)
Daniele Torella, Georgina M Ellison, Michele Torella, Carla Vicinanza, Iolanda Aquila, Claudio Iaconetti, Mariangela Scalise, Fabiola Marino, Beverley J Henning, Fiona C Lewis, Clarice Gareri, Nadia Lascar, Giovanni Cuda, Teresa Salvatore, Gianantonio Nappi, Ciro Indolfi, Roberto Torella, Domenico Cozzolino, Ferdinando Carlo Sasso
ABSTRACT
Diabetes mellitus (DM) has multifactorial detrimental effects on myocardial tissue. Recently, carbonic anhydrases (CAs) have been shown to play a major role in diabetic microangiopathy but their role in the diabetic cardiomyopathy is still unknown. We obtained left ventricular samples from patients with DM type 2 (DM-T2) and nondiabetic (NDM) patients with postinfarct heart failure who were undergoing surgical coronary revascularization. Myocardial levels of CA-I and CA-II were 6- and 11-fold higher, respectively, in DM-T2 versus NDM patients. Elevated CA-I expression was mainly localized in the cardiac interstitium and endothelial cells. CA-I induced by high glucose levels hampers endothelial cell permeability and determines endothelial cell apoptosis in vitro. Accordingly, capillary density was significantly lower in the DM-T2 myocardial samples (mean±SE=2152±146 versus 4545±211/mm(2)). On the other hand, CA-II was mainly upregulated in cardiomyocytes. The latter was associated with sodium-hydrogen exchanger-1 hyperphosphorylation, exaggerated myocyte hypertrophy (cross-sectional area 565±34 versus 412±27 μm(2)), and apoptotic death (830±54 versus 470±34 per 10(6) myocytes) in DM-T2 versus NDM patients. CA-II is activated by high glucose levels and directly induces cardiomyocyte hypertrophy and death in vitro, which are prevented by sodium-hydrogen exchanger-1 inhibition. CA-II was shown to be a direct target for repression by microRNA-23b, which was downregulated in myocardial samples from DM-T2 patients. MicroRNA-23b is regulated by p38 mitogen-activated protein kinase, and it modulates high-glucose CA-II-dependent effects on cardiomyocyte survival in vitro. Myocardial CA activation is significantly elevated in human diabetic ischemic cardiomyopathy. These data may open new avenues for targeted treatment of diabetic heart failure.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human CA2
Sigma-Aldrich
KR-32568, ≥98% (HPLC), solid