Merck
  • [Fe(CN)5(isoniazid)](3-): an iron isoniazid complex with redox behavior implicated in tuberculosis therapy.
[Fe(CN)5(isoniazid)](3-): an iron isoniazid complex with redox behavior implicated in tuberculosis therapy.
Journal of inorganic biochemistry (2014-09-06)
Eduardo Henrique Silva Sousa, Francisca Gilmara de Mesquita Vieira, Jennifer S Butler, Luiz Augusto Basso, Diógenes S Santiago, Izaura C N Diógenes, Luiz Gonzaga de França Lopes, Peter J Sadler
ABSTRACT
Tuberculosis has re-emerged as a worldwide threat, which has motivated the development of new drugs. The antituberculosis complex Na3[Fe(CN)5(isoniazid)] (IQG607) in particular is of interest on account of its ability to overcome resistance. IQG607 has the potential for redox-mediated-activation, in which an acylpyridine (isonicotinoyl) radical could be generated without assistance from the mycobacterial KatG enzyme. Here, we have investigated the reactivity of IQG607 toward hydrogen peroxide and superoxide, well-known intracellular oxidizing agents that could play a key role in the redox-mediated-activation of this compound. HPLC, NMR and electronic spectroscopy studies showed a very fast oxidation rate for bound isoniazid, over 460-fold faster than free isoniazid oxidation. A series of EPR spin traps were used for detection of isonicotinoyl and derived radicals bound to iron. This is the first report for an isonicotinoyl radical bound to a metal complex, supported by (14)N and (1)H hyperfine splittings for the POBN and PBN trapped radicals. POBN and PBN exhibited average hyperfine coupling constants of aN=15.6, aH=2.8 and aN=15.4, aH=4.7, respectively, which are in close agreement to the isonicotinoyl radical. Radical generation is thought to play a major role in the mechanism of action of isoniazid and this work provides strong evidence for its production within IQG607, which, along with biological and chemical oxidation data, support a redox-mediated activation mechanism. More generally the concept of redox activation of metallo prodrugs could be applied more widely for the design of therapeutic agents with novel mechanisms of action.
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
α-(4-Pyridyl N-oxide)-N-tert-butylnitrone, 99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Isonicotinamide, ReagentPlus®, 99%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
α-(4-Pyridyl N-oxide)-N-tert-butylnitrone, ~95%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland