Merck
  • Enlarging the toolbox for allergen epitope definition with an allergen-type model protein.
Enlarging the toolbox for allergen epitope definition with an allergen-type model protein.
PloS one (2014-10-31)
Hanna Berkner, Christian Seutter von Loetzen, Maximilian Hartl, Stefanie Randow, Michaela Gubesch, Lothar Vogel, Felix Husslik, Andreas Reuter, Jonas Lidholm, Barbara Ballmer-Weber, Stefan Vieths, Paul Rösch, Dirk Schiller
ABSTRACT
Birch pollen-allergic subjects produce polyclonal cross-reactive IgE antibodies that mediate pollen-associated food allergies. The major allergen Bet v 1 and its homologs in plant foods bind IgE in their native protein conformation. Information on location, number and clinical relevance of IgE epitopes is limited. We addressed the use of an allergen-related protein model to identify amino acids critical for IgE binding of PR-10 allergens. Norcoclaurine synthase (NCS) from meadow rue is structurally homologous to Bet v 1 but does not bind Bet v 1-reactive IgE. NCS was used as the template for epitope grafting. NCS variants were tested with sera from 70 birch pollen allergic subjects and with monoclonal antibody BV16 reported to compete with IgE binding to Bet v 1. We generated an NCS variant (Δ29NCSN57/I58E/D60N/V63P/D68K) harboring an IgE epitope of Bet v 1. Bet v 1-type protein folding of the NCS variant was evaluated by 1H-15N-HSQC NMR spectroscopy. BV16 bound the NCS variant and 71% (50/70 sera) of our study population showed significant IgE binding. We observed IgE and BV16 cross-reactivity to the epitope presented by the NCS variant in a subgroup of Bet v 1-related allergens. Moreover BV16 blocked IgE binding to the NCS variant. Antibody cross-reactivity depended on a defined orientation of amino acids within the Bet v 1-type conformation. Our system allows the evaluation of patient-specific epitope profiles and will facilitate both the identification of clinically relevant epitopes as biomarkers and the monitoring of therapeutic outcomes to improve diagnosis, prognosis, and therapy of allergies caused by PR-10 proteins.
Sigma-Aldrich
Anti-Mouse IgG (γ-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)