Merck
  • Matrix effects break the LC behavior rule for analytes in LC-MS/MS analysis of biological samples.
Matrix effects break the LC behavior rule for analytes in LC-MS/MS analysis of biological samples.
Experimental biology and medicine (Maywood, N.J.) (2014-10-12)
Nianbai Fang, Shanggong Yu, Martin Jj Ronis, Thomas M Badger
ABSTRACT
High-performance liquid chromatography (HPLC) and liquid chromatography-tandem mass spectrometry (LC-MS/MS) are generally accepted as the preferred techniques for detecting and quantitating analytes of interest in biological matrices on the basis of the rule that one chemical compound yields one LC-peak with reliable retention time (Rt.). However, in the current study, we have found that under the same LC-MS conditions, the Rt. and shape of LC-peaks of bile acids in urine samples from animals fed dissimilar diets differed significantly among each other. To verify this matrix effect, 17 authentic bile acid standards were dissolved in pure methanol or in methanol containing extracts of urine from pigs consuming either breast milk or infant formula and analyzed by LC-MS/MS. The matrix components in urine from piglets fed formula significantly reduced the LC-peak Rt. and areas of bile acids. This is the first characterization of this matrix effect on Rt. in the literature. Moreover, the matrix effect resulted in an unexpected LC behavior: one single compound yielded two LC-peaks, which broke the rule of one LC-peak for one compound. The three bile acid standards which exhibited this unconventional LC behavior were chenodeoxycholic acid, deoxycholic acid, and glycocholic acid. One possible explanation for this effect is that some matrix components may have loosely bonded to analytes, which changed the time analytes were retained on a chromatography column and interfered with the ionization of analytes in the MS ion source to alter the peak area. This study indicates that a comprehensive understanding of matrix effects is needed towards improving the use of HPLC and LC-MS/MS techniques for qualitative and quantitative analyses of analytes in pharmacokinetics, proteomics/metabolomics, drug development, and sports drug testing, especially when LC-MS/MS data are analyzed by automation software where identification of an analyte is based on its exact molecular weight and Rt.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Supelco
Methanol, analytical standard
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Ursodeoxycholic acid, ≥99%
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Lithocholic acid, ≥95%
Sigma-Aldrich
Chenodeoxycholic acid
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Ursodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard
Lithocholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)