Merck
  • Microextraction of non-steroidal anti-inflammatory drugs from waste water samples by rotating-disk sorptive extraction.
Microextraction of non-steroidal anti-inflammatory drugs from waste water samples by rotating-disk sorptive extraction.
Talanta (2014-07-26)
Valentina Manzo, Luis Honda, Orielle Navarro, Loreto Ascar, Pablo Richter
ABSTRACT
In this study, six non-steroidal anti-inflammatory drugs (NSAIDs) were extracted from water samples using the rotating-disk sorptive extraction (RDSE) technique. The extraction disk device contains a central cavity that allows for the incorporation of a powdered sorbent phase (Oasis™ HLB). The analytes were extracted from water and pre-concentrated on the sorbent to reach the extraction equilibrium, and then they were desorbed with solvent, derivatized and determined by gas chromatography-mass spectrometry (GC-MS). The variables for the extraction were studied using high performance liquid chromatography with a diode array detector (HPLC-DAD) to avoid the derivatization step, and the optimum values were as follows: 60 mg of Oasis™ HLB, a rotation velocity of 3,000 rpm, a pH of 2, a sample volume of 50 mL, and an extraction time of approximately 90-100 min. The recoveries ranged from 71 to 104%, with relative standard deviations (RSD) between 2 and 8%. The detection limits ranged from 0.001 to 0.033 µg L(-1). The described method was applied to the analysis of influents and effluents from wastewater treatment plants (WWTP) in Santiago, Chile. The concentrations of the detected drugs ranged from 1.5 to 13.4 µg L(-1) and from 1.0 to 3.2 µg L(-1) in the influents and effluents, respectively. The samples were extracted by solid phase extraction (SPE). No significant differences were observed in the determined concentrations for most of the NSAIDs, indicating that RDSE is an alternative method for the preparation of water samples.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Ibuprofen
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%