Merck
  • Autocrine/paracrine function of globular adiponectin: inhibition of lipid metabolism and inflammatory response in 3T3-L1 adipocytes.
Autocrine/paracrine function of globular adiponectin: inhibition of lipid metabolism and inflammatory response in 3T3-L1 adipocytes.
Journal of cellular biochemistry (2014-12-11)
Yulia Lazra, Alona Falach, Lital Frenkel, Konstantin Rozenberg, Sanford Sampson, Tovit Rosenzweig
ABSTRACT
Adiponectin is an adipose-derived hormone, with beneficial effects on insulin sensitivity and inflammation. The aim of this study was to clarify the autocrine/paracrine effects of globular adiponectin (gAd) administrated during differentiation on the function of the mature adipocytes. Experiments were performed on 3T3-L1 preadipocytes treated with gAd (10 nM), starting at an early stage of differentiation. gAd treatment during differentiation was without effect on mRNA expression of adiponectin and AdipoR2, but increased AdipoR1 expression. PPARgamma, perillipin and FABP4 mRNA expressions were lower in gAd-treated adipocytes, accompanied by a reduction in lipid accumulation. While mRNA expression of HSL was not affected by gAd compared to untreated adipocytes, both ATGL and FAS were reduced, indicating that gAd regulates both lipolysis and lipogenesis. PPARα, ACOX2 and UCPs mRNA expressions were not affected by gAd, indicating that the reduction in lipid content is not attributed to an increase in fatty-acid oxidation. In accord with the lower PPARγ expression, there was reduced Glut4 mRNA expression; however, insulin-induced PKB phosphorylation was enhanced by gAd, and glucose uptake was not altered. To investigate the effect of gAd on LPS-induced inflammatory response, cells were treated with gAd either during differentiation or 24 h before induction of inflammation in differentiated adipocytes. LPS-induced inflammatory response, as indicated by increase in IL6 and MCP-1 mRNA expression. gAd given through differentiation was much more effective in abrogating LPS-dependent cytokines production than gAd given to the mature adipocyte. In conclusion, elevated gAd at differentiation of 3T3-L1 cells leads to reduced lipid content, reduced lipid metabolism and high resistance to inflammation.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), BioXtra
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%