Merck
  • Elongation factor-2 kinase regulates TG2/β1 integrin/Src/uPAR pathway and epithelial-mesenchymal transition mediating pancreatic cancer cells invasion.
Elongation factor-2 kinase regulates TG2/β1 integrin/Src/uPAR pathway and epithelial-mesenchymal transition mediating pancreatic cancer cells invasion.
Journal of cellular and molecular medicine (2014-09-13)
Ahmed A Ashour, Nilgun Gurbuz, Sultan Neslihan Alpay, Abdel-Aziz H Abdel-Aziz, Ahmed M Mansour, Longfei Huo, Bulent Ozpolat
ABSTRACT
Pancreatic ductal adenocarcinoma is one of the lethal cancers with extensive local tumour invasion, metastasis, early systemic dissemination and poorest prognosis. Thus, understanding the mechanisms regulating invasion/metastasis and epithelial-mesenchymal transition (EMT), is the key for developing effective therapeutic strategies for pancreatic cancer (PaCa). Eukaryotic elongation factor-2 kinase (eEF-2K) is an atypical kinase that we found to be highly up-regulated in PaCa cells. However, its role in PaCa invasion/progression remains unknown. Here, we investigated the role of eEF-2K in cellular invasion, and we found that down-regulation of eEF-2K, by siRNA or rottlerin, displays impairment of PaCa cells invasion/migration, with significant decreases in the expression of tissue transglutaminase (TG2), the multifunctional enzyme implicated in regulation of cell attachment, motility and survival. These events were associated with reductions in β1 integrin/uPAR/MMP-2 expressions as well as decrease in Src activity. Furthermore, inhibition of eEF-2K/TG2 axis suppresses the EMT, as demonstrated by the modulation of the zinc finger transcription factors, ZEB1/Snail, and the tight junction proteins, claudins. Importantly, while eEF-2K silencing recapitulates the rottlerin-induced inhibition of invasion and correlated events, eEF-2K overexpression, by lentivirus-based expression system, suppresses such rottlerin effects and potentiates PaCa cells invasion/migration capability. Collectively, our results show, for the first time, that eEF-2K is involved in regulation of the invasive phenotype of PaCa cells through promoting a new signalling pathway, which is mediated by TG2/β1 integrin/Src/uPAR/MMP-2, and the induction of EMT biomarkers which enhance cancer cell motility and metastatic potential. Thus, eEF-2K could represent a novel potential therapeutic target in pancreatic cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Eef2k
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tgm2
Sigma-Aldrich
MISSION® esiRNA, targeting human TGM2
Sigma-Aldrich
MISSION® esiRNA, targeting human EEF2K
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Src