Merck
  • Decreased expression of the long non-coding RNA FENDRR is associated with poor prognosis in gastric cancer and FENDRR regulates gastric cancer cell metastasis by affecting fibronectin1 expression.
Decreased expression of the long non-coding RNA FENDRR is associated with poor prognosis in gastric cancer and FENDRR regulates gastric cancer cell metastasis by affecting fibronectin1 expression.
Journal of hematology & oncology (2014-08-30)
Tong-Peng Xu, Ming-de Huang, Rui Xia, Xin-Xin Liu, Ming Sun, Li Yin, Wen-Ming Chen, Liang Han, Er-Bao Zhang, Rong Kong, Wei De, Yong-Qian Shu
ABSTRACT
FENDRR is a long non-coding RNAs (lncRNA) that binds to polycomb repressive complexe 2 (PRC2) to epigenetically regulate the expression of its target gene. The clinical role of FENDRR in carcinomas remains yet to be found. Real-time polymerase chain reaction (PCR) was used to examine FENDRR expression in gastric cancer cell lines/tissues compared with normal epithelial cells/adjacent non-tumorous tissues. Cell proliferation assays, Wound healing assays, and in vitro and in vivo invasion and migration assays were performed to detect the biological effects of FENDRR in gastric cancer cells. Real-time PCR, western-blot and immunohistochemistry were used to evaluate the mRNA and protein expression of fibronectin1 (FN1). Secreted matrix metalloproteinase (MMP) activities were detected and characterized using gelatin zymography assay. FENDRR was downregulated in gastric cancer cell lines and cancerous tissues, as compared with normal gastric epithelial cells and adjacent noncancerous tissue samples. Low FENDRR expression was correlated with deeper tumor invasion (p < 0.001), higher tumor stage (p = 0.001), and lymphatic metastasis (p = 0.007). Univariate and multivariate analyses indicated that low FENDRR expression predicted poor prognosis. Histone deacetylation was involved in the downregulation of FENDRR in gastric cancer cells. FENDER overexpression suppressed invasion and migration by gastric cancer cells in vitro, by downregulating FN1 and MMP2/MMP9 expression. Low expression of the lncRNA FENDRR occurs in gastric cancer and is associated with poor prognosis. Thus, FENDRR plays an important role in the progression and metastasis of gastric cancer.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
MISSION® esiRNA, targeting human FN1
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fn1
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard