Merck
  • Homogeneous detection of caspase-3 using intrinsic fluorescence resonance energy transfer (iFRET).
Homogeneous detection of caspase-3 using intrinsic fluorescence resonance energy transfer (iFRET).
Biosensors & bioelectronics (2014-09-15)
Hyo Jin Kang, Ju Hwan Kim, Sang J Chung
ABSTRACT
Caspase-3 is an apoptotic cysteine protease and its aberrancy is highly implicated to numerous diseases thereby rendering caspase-3 activity as an important disease marker. Most caspase-3 sensors are caspase-3 substrates of which the fluorescence signals are turned on upon catalytic cleavage by active caspase-3. However, once the signal is generated, the fluorescence does not disappear albeit caspase-3 activity is abolished. Recently, we and other groups have developed the intrinsic Förster resonance energy transfer (iFRET) technique, which utilizes tryptophan residues of the target proteins and target-specific probes, as FRET donors and acceptors, respectively. Due to this principle, iFRET does not require the labeling of target proteins. In this work, we report the development of caspase-3 specific iFRET probes by structure-based design and synthesis, and the successful detection of caspase-3 in cell lysates as well as in its purified form. The limit-of-detection (LOD) of the probes in case of purified caspase-3 was found to be 1.4-1.5 nM. The designed probes did not bind to either procaspase-3 or C163S caspase-3, which are catalytic inactive, confirming that the observed iFRET signal correlates to the catalytic activity of caspase-3. Furthermore, in competition experiments with Ac-DEVD-CHO, a known competitive inhibitor of caspase-3, the iFRET signal was inhibited.
Product Description
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrabutylammonium fluoride on silica gel, ~1.5 mmol/g (F-)
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SAFC
HEPES
Trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%