Merck
  • Effects of selective COX-2 inhibition on allergen-induced bronchoconstriction and airway inflammation in asthma.
Effects of selective COX-2 inhibition on allergen-induced bronchoconstriction and airway inflammation in asthma.
The Journal of allergy and clinical immunology (2014-01-28)
Kameran Daham, Anna James, David Balgoma, Maciej Kupczyk, Bo Billing, Agneta Lindeberg, Elisabeth Henriksson, Garret A FitzGerald, Craig E Wheelock, Sven-Erik Dahlén, Barbro Dahlén
ABSTRACT
Prostaglandins that constrict and relax airways are synthesized in reactions catalyzed by either COX-1 or COX-2. It is not known whether selective inhibition of COX-2 makes asthmatic responses better or worse. To determine the effects of the selective COX-2 inhibitor, etoricoxib, on allergen-induced bronchoconstriction in asthmatic subjects. Sixteen subjects with mild atopic asthma underwent rising dose inhalation challenges with allergen or methacholine to determine PD20 FEV1 during a control study period or after 10 to 13 days of treatment with etoricoxib (90 mg once daily). The order of study periods was randomized with at least 2-week washout periods. Induced sputum cells and fractional exhaled nitric oxide levels were used to assess airway inflammation and blood assays for COX-1 and COX-2 activity to assess enzyme inhibition. Urinary excretion of lipids was used to assess prostaglandin biosynthesis. Etoricoxib did not change baseline lung function, nor airway responsiveness to allergen or to methacholine. Neither were the allergen-induced increase in sputum eosinophils and fractional exhaled nitric oxide levels affected by treatment. The biochemical effectiveness of the treatment was established both in the blood assays and by the distinct inhibitory effect of etoricoxib on urinary excretion of tetranor-prostaglandin E2 (P < .001). This first study of COX-2 inhibition in provoked asthma found no negative effects of etoricoxib on allergen-induced airflow obstruction and sputum eosinophils, basal lung function, or methacholine responsiveness. The study suggests that short-term use of COX-2 inhibitors is safe in subjects with asthma.