Merck
  • Safety and tolerability of intracerebroventricular PDGF-BB in Parkinson's disease patients.
Safety and tolerability of intracerebroventricular PDGF-BB in Parkinson's disease patients.
The Journal of clinical investigation (2015-02-18)
Gesine Paul, Olof Zachrisson, Andrea Varrone, Per Almqvist, Markus Jerling, Göran Lind, Stig Rehncrona, Bengt Linderoth, Hjalmar Bjartmarz, Lisa L Shafer, Robert Coffey, Mikael Svensson, Katarina Jansson Mercer, Anton Forsberg, Christer Halldin, Per Svenningsson, Håkan Widner, Jonas Frisén, Sven Pålhagen, Anders Haegerstrand
ABSTRACT
BACKGROUND. Recombinant human PDGF-BB (rhPDGF-BB) reduces Parkinsonian symptoms and increases dopamine transporter (DAT) binding in several animal models of Parkinson's disease (PD). Effects of rhPDGF-BB are the result of proliferation of ventricular wall progenitor cells and reversed by blocking mitosis. Based on these restorative effects, we assessed the safety and tolerability of intracerebroventricular (i.c.v.) rhPDGF-BB administration in individuals with PD. METHODS. We conducted a double-blind, randomized, placebo-controlled phase I/IIa study at two clinical centers in Sweden. Twelve patients with moderate PD received rhPDGF-BB via an implanted drug infusion pump and an investigational i.c.v. catheter. Patients were assigned to a dose cohort (0.2, 1.5, or 5 μg rhPDGF-BB per day) and then randomized to active treatment or placebo (3:1) for a 12-day treatment period. The primary objective was to assess safety and tolerability of i.c.v.-delivered rhPDGF-BB. Secondary outcome assessments included several clinical rating scales and changes in DAT binding. The follow-up period was 85 days. RESULTS. All patients completed the study. There were no unresolved adverse events. Serious adverse events occurred in three patients; however, these were unrelated to rhPDGF-BB administration. Secondary outcome parameters did not show dose-dependent changes in clinical rating scales, but there was a positive effect on DAT binding in the right putamen. CONCLUSION. At all doses tested, i.c.v. administration of rhPDGF-BB was well tolerated. Results support further clinical development of rhPDGF-BB for patients with PD. TRIAL REGISTRATION. Clinical Trials.gov NCT00866502. FUNDING. Newron Sweden AB (former NeuroNova AB) and Swedish Governmental Agency for Innovation Systems (VINNOVA).
Sigma-Aldrich
Anti-PDGF-BB Antibody, from rabbit, purified by affinity chromatography
Tantalum(V) ethoxide, packaged for use in deposition systems
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis