Merck
  • Fluorescent carbon nanodots for sensitive and selective detection of tannic acid in wines.
Fluorescent carbon nanodots for sensitive and selective detection of tannic acid in wines.
Talanta (2014-12-06)
Gaber Hashem Gaber Ahmed, Rosana Badía Laíño, Josefa Angela García Calzón, Marta Elena Díaz García
ABSTRACT
Herein we describe an easy one step synthesis of carbon nanodots (C-dots) by thermal carbonization of 6-bromohexylboronic acid using two different amine compounds, polyethyleneglycol bis(3-aminopropyl (PEGA) and 1,2-aminopropane (DPA), at 180 °C in atmospheric oxygen. The as-synthesized C-dots were characterized by FTIR, HRTEM, NMR and fluorescence. The C-dots prepared using PEGA showed a strong emission at 440 nm with excitation at 362 nm. These C-dots exhibited analytical potential as sensing probes for tannic acid (TA) determination. pH effect, interferences, and analytical performance of the method were investigated. The method was found effective in the linear concentration range from 0.1 to 10 mg L(-1) TA achieving a limit of detection equal 0.018 mg L(-1) TA. The applicability of the method was demonstrated by direct measurements of TA in red and white wine samples. Validation of the method was achieved by spiking the wine samples with different standard TA concentrations obtaining recoveries in the range (90-112.5%). A probable mechanism by which TA quenched the C-dots fluorescence was proposed.
Product Description
USP
Fructose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
L-Ascorbic acid, analytical standard
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Fructose, European Pharmacopoeia (EP) Reference Standard
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium sulfite, BioXtra, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%