Merck
  • Toll-like receptor 4 ablation in mdx mice reveals innate immunity as a therapeutic target in Duchenne muscular dystrophy.
Toll-like receptor 4 ablation in mdx mice reveals innate immunity as a therapeutic target in Duchenne muscular dystrophy.
Human molecular genetics (2015-01-02)
Christian Giordano, Kamalika Mojumdar, Feng Liang, Christian Lemaire, Tong Li, John Richardson, Maziar Divangahi, Salman Qureshi, Basil J Petrof
ABSTRACT
Toll-like receptor 4 (TLR4) recognizes specific structural motifs associated with microbial pathogens and also responds to certain endogenous host molecules associated with tissue damage. In Duchenne muscular dystrophy (DMD), inflammation plays an important role in determining the ultimate fate of dystrophic muscle fibers. In this study, we used TLR4-deficient dystrophic mdx mice to assess the role of TLR4 in the pathogenesis of DMD. TLR4 expression was increased and showed enhanced activation following agonist stimulation in mdx diaphragm muscle. Genetic ablation of TLR4 led to significantly increased muscle force generation in dystrophic diaphragm muscle, which was associated with improved histopathology including decreased fibrosis, as well as reduced pro-inflammatory gene expression and macrophage infiltration. TLR4 ablation in mdx mice also altered the phenotype of muscle macrophages by inducing a shift toward a more anti-inflammatory (iNOS(neg) CD206(pos)) profile. In vitro experiments confirmed that lack of TLR4 is sufficient to influence macrophage activation status in response to classical polarizing stimuli such as IFN-gamma and IL-4. Finally, treatment of dystrophic mice with glycyrrhizin, an inhibitor of the endogenous TLR4 ligand, high mobility group box (HMGB1), also pointed to involvement of the HMGB1-TLR4 axis in promoting dystrophic diaphragm pathology. Taken together, our findings reveal TLR4 and the innate immune system as important players in the pathophysiology of DMD. Accordingly, targeting either TLR4 or its endogenous ligands may provide a new therapeutic strategy to slow disease progression.
Product Description
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥95.0% (NT)
Supelco
Glycyrrhizic acid ammonium salt, analytical standard, suitable for HPLC
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Ponceau S solution, BioReagent, suitable (for use in cellulose acetate electrophoresis), 0.1 % (w/v) in 5% acetic acid
Supelco
2-Methylbutane, analytical standard
Ammonium glycyrrhizate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
2-Methylbutane, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥70% (HPLC)
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Glycyrrhizate monoammonium, European Pharmacopoeia (EP) Reference Standard