Merck
  • Separation and characterisation of detonation nanodiamond by capillary zone electrophoresis.
Separation and characterisation of detonation nanodiamond by capillary zone electrophoresis.
Electrophoresis (2014-03-22)
Emer Duffy, Dimitar P Mitev, Pavel N Nesterenko, Artaches A Kazarian, Brett Paull
ABSTRACT
A new method for the characterisation of purified detonation nanodiamond (DND) using CZE has been developed. The influence of BGE conditions on electrophoretic mobility, peak shape and particle aggregation was investigated, with resultant observations supported by zeta potential approximations and particle size measurements. Sodium tetraborate (pH 9.3), Tris (pH 9.3) and sodium phosphate (pH 7) were used in studying the BGE concentration effect on a commercial source of chemically stabilised DND. The BGE concentration had a strong effect on the stability of DND in suspension. The formation of aggregates of various sizes was observed as BGE concentration increased. The effect of pH on the electromigration of DND was examined using sodium phosphate (pH 8 and 10). The CZE method was subsequently applied to four different DND samples, which had undergone different routes of purification following detonation synthesis. Each sample produced a unique electrophoretic peak or profile in sodium tetraborate buffer (pH 9.3), such that the actual separation of DND samples from different sources could be achieved.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O