Merck
  • Genome-wide identification of long noncoding RNAs in rat models of cardiovascular and renal disease.
Genome-wide identification of long noncoding RNAs in rat models of cardiovascular and renal disease.
Hypertension (Dallas, Tex. : 1979) (2014-11-12)
Kathirvel Gopalakrishnan, Sivarajan Kumarasamy, Blair Mell, Bina Joe
ABSTRACT
Long noncoding RNAs (lncRNAs) are an emerging class of genomic regulatory molecules reported in various species. In the rat, which is one of the major mammalian model organisms, discovery of lncRNAs on a genome-wide scale is lagging. Renal lncRNA sequencing and lncRNA transcriptome analysis were conducted in 3 rat strains that are widely used in cardiovascular and renal research: the Dahl salt-sensitive rat, the spontaneously hypertensive rat, and the Dahl salt-resistant rat. Through the RNA sequencing approach, 3273 transcripts were identified as rat lncRNAs. A majority of lncRNAs were without predicted target genes. Differential expression of 273 and 749 lncRNAs was detected between Dahl salt-sensitive versus Dahl salt-resistant and Dahl salt-sensitive versus spontaneously hypertensive rat comparisons, respectively. To couple the observed differential expression of lncRNAs with the status of mRNAs, an mRNA transcriptome analysis was conducted. Several cis mRNA genes were coregulated with lncRNAs. Of these, the protein expression status of 4 target genes, Asb3, Chac2, Pex11b, and Sp5, were differentially expressed between the relevant strain comparisons, thereby suggesting that the differentially expressed lncRNAs associated with these genes are candidate genetic determinants of blood pressure. This study serves as a first-generation catalog of rat lncRNAs and illustrates the prioritization of lncRNAs as candidates for complex polygenic traits.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)