Merck
  • pH-sensitive nanoformulated triptolide as a targeted therapeutic strategy for hepatocellular carcinoma.
pH-sensitive nanoformulated triptolide as a targeted therapeutic strategy for hepatocellular carcinoma.
ACS nano (2014-08-06)
Daishun Ling, Hongping Xia, Wooram Park, Michael J Hackett, Changyeong Song, Kun Na, Kam Man Hui, Taeghwan Hyeon
ABSTRACT
Hepatocellular carcinoma (HCC) has one of the worst prognoses for survival as it is poorly responsive to both conventional chemotherapy and mechanism-directed therapy. This results from a lack of therapeutic concentration in the tumor tissue coupled with the highly toxic off-site effects exhibited by these compounds. Consequently, we believe the best packaging for holistic therapy for HCC will involve three components: a potent therapeutic, a rationally designed drug delivery vehicle to enrich the target site concentration of the drug, and a surface ligand that can enable a greater propensity to internalization by tumor cells compared to the parenchyma. We screened a library containing hundreds of compounds against a panel of HCC cells and found the natural product, triptolide, to be more effective than sorafenib, doxorubicin, and daunorubicin, which are the current standards of therapy. However, the potential clinical application of triptolide is limited due to its poor solubility and high toxicity. Consequently, we synthesized tumor pH-sensitive nanoformulated triptolide coated with folate for use in an HCC-subpopulation that overexpresses the folate receptor. Our results show triptolide itself can prevent disease progression, but at the cost of significant toxicity. Conversely, our pH-sensitive nanoformulated triptolide facilitates uptake into the tumor, and specifically tumor cells, leading to a further increase in efficacy while mitigating systemic toxicity.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Octadecylamine, technical grade, 90%
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Octadecylamine, ≥99.0% (GC)
Sigma-Aldrich
Bis(trichloromethyl) carbonate, purum, ≥99.0% (AT)
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%