Merck
  • Analysis of the essentiality of ROM2 genes in the pathogenic yeasts Candida glabrata and Candida albicans using temperature-sensitive mutants.
Analysis of the essentiality of ROM2 genes in the pathogenic yeasts Candida glabrata and Candida albicans using temperature-sensitive mutants.
Journal of applied microbiology (2015-01-22)
T Kanno, D Takekawa, Y Miyakawa
ABSTRACT
To analyse the essentiality of the ROM2 genes originating from the pathogenic yeasts Candida glabrata and Candida albicans by using temperature-sensitive (ts) mutants. Based on the general concepts that ts mutations are generated by virtue of point mutation within essential genes, we have previously established a novel method (termed 'ETS system' for screening and identification of essential genes using ts mutants of C. glabrata). According to this ETS system, the present study successfully identified a putative C. glabrata ROM2 homologue as an essential gene that complements its point mutation (Cys-1275/Tyr substitution). The C. albicans ROM2 mutant (Cys-1281/Tyr), constructed patterned after this point mutation, also displayed ts phenotype. Both ts mutants recovered colony-forming ability, with concomitant suppression of lysis phenotype, at the elevated temperature in the presence of 1 mol l(-1) sorbitol as an osmotic stabilizer. Sequence alignment revealed that human genome possesses relatively low homology against Rom2 homologues, which are highly conserved among yeast species. ROM2 genes of C. glabrata and C. albicans are essential for viability, probably involved in cell wall integrity. ROM2 genes essential for both Candida species may be a potentially useful antifungal targets from chemotherapeutic viewpoint.
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides