Merck
  • Developmental exposure to the organophosphorus flame retardant tris(1,3-dichloro-2-propyl) phosphate: estrogenic activity, endocrine disruption and reproductive effects on zebrafish.
Developmental exposure to the organophosphorus flame retardant tris(1,3-dichloro-2-propyl) phosphate: estrogenic activity, endocrine disruption and reproductive effects on zebrafish.
Aquatic toxicology (Amsterdam, Netherlands) (2015-02-01)
Qiangwei Wang, James C W Lam, Jian Han, Xianfeng Wang, Yongyong Guo, Paul K S Lam, Bingsheng Zhou
ABSTRACT
Tris(1,3-dichloro-2-propyl) phosphate (TDCPP) is an organophosphate flame retardant that is detectable in the environment and biota, prompting concern over its risk to wildlife and human health. Our objective was to investigate whether long-term exposure to low concentrations of TDCPP can affect fish reproduction. Zebrafish embryos were exposed to low concentrations (0, 4, 20 and 100μg/L) of TDCPP from 2h post-fertilization until sexual maturation. Exposure to TDCPP significantly increased plasma estradiol and testosterone levels in females, but had no effect in males. TDCPP exposure also caused a significant reduction in fecundity as indicated by decreased egg production. Real-time PCR was performed to examine selected genes in the hypothalamic-pituitary-gonadal (HPG) axis and liver. Principle component analysis (PCA) showed that sex hormone levels and fecundity were related to the mRNA level of several genes in the HPG axis. Furthermore, hepatic vitellogenin (vtg1 and vtg3) expression was upregulated in both females and males, suggesting TDCPP has estrogenic activity. Histological examination revealed promotion of oocyte maturation in the females, but retardation of spermiation in males. Reduced egg quality (e.g., egg diameter) and increased malformation rates were observed in the F1 generation. Chemical analysis showed significant levels of TDCPP and its metabolite bis(1,3-dichloro-2-propyl) phosphate in the gonads of males and females. In conclusion, long-term exposure to low concentrations of TDCPP impairs fish reproduction.
Product Description
Supelco
TDCPP, PESTANAL®, analytical standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Tripropyl phosphate, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Trimethyl phosphate, ≥99%
Sigma-Aldrich
Trimethyl phosphate, 97%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)