Merck
  • Superficially porous particles vs. fully porous particles for bonded high performance liquid chromatographic chiral stationary phases: isopropyl cyclofructan 6.
Superficially porous particles vs. fully porous particles for bonded high performance liquid chromatographic chiral stationary phases: isopropyl cyclofructan 6.
Journal of chromatography. A (2014-08-30)
Daniel A Spudeit, Maressa D Dolzan, Zachary S Breitbach, William E Barber, Gustavo A Micke, Daniel W Armstrong
ABSTRACT
This work reports a comparison of HPLC separations of enantiomers with chiral stationary phases (CSPs) prepared by chemically bonding cyclofructan-6, functionalized with isopropyl carbamate groups on fully and superficially porous particles (SPPs). The chromatographic performance of the superficially porous CSP based column was compared with columns packed with 5 μm and 3 μm fully porous particles (FPPs). At a flow rate of 3.0 mL/min the number of plates on column afforded by the SPP column was ∼7× greater than the number of plates on column (same length) obtained when using the 5 μm FPP based column. The flow rate providing the highest efficiency separation was ∼1.0 mL/min for the SPP column while it was ∼0.5 mL/min for both FPP columns. It was found that the selectivity and resolution of the separations were comparable between fully porous and superficially porous based columns (under constant mobile phase conditions), even though the SPP column contained lower absolute amounts of chiral selector. When tested under constant retention conditions, the SPP based CSP greatly improved resolution compared to the FPP based columns. At high flow rates the efficiency gained by using superficially porous CSP was accentuated. The advantages of columns based on SPPs become more obvious from the viewpoint of plate numbers and resolution per analysis time.
Product Description
Supelco
Acetonitrile, analytical standard
Supelco
Pyridine, analytical standard
Supelco
Toluene, analytical standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Pyridine, ≥99%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard