Merck
  • An "ex vivo" model to evaluate toxicological responses to mixtures of contaminants in cetaceans: integumentum biopsy slices.
An "ex vivo" model to evaluate toxicological responses to mixtures of contaminants in cetaceans: integumentum biopsy slices.
Environmental toxicology (2013-01-23)
Maria Cristina Fossi, Silvia Casini, Silvia Maltese, Cristina Panti, Giacomo Spinsanti, Letizia Marsili
ABSTRACT
The need for powerful new tools to detect the effects of chemical pollution, in particular of persistent organic pollutants (POPs) and polycyclic aromatic hydrocarbons (PAHs) on Mediterranean cetaceans led us to develop and apply a suite of sensitive biomarkers for integument biopsies of stranded and free-ranging animals. This multi-response ex vivo method has the aim to detect toxicological effects of contaminant mixtures. In the present study, we applied an ex vivo assay using skin biopsy and liver slices, combining molecular biomarkers [Western blot of Cytochrome P450 1A1 (CYP1A1) and Cytochrome P450 2B (CYP2B)] and gene expression biomarkers (Quantitative real-time PCR of CYP1A1, heat shock protein 70, estrogen receptor alpha and E2F transcription factor) in response to chemical exposure [organochlorines compounds (OCs), polybrominated diphenyl ethers (PBDEs), and PAHs] for stranded Mediterranean Stenella coeruleoalba. The main goal of this experiment was to identify the biomarker and/or a suite of biomarkers that could best detect the presence of a specific class of pollutants (OCs, PBDEs, and PAHs) or a mixture of them. This multi-response biomarker methodology revealed an high sensitivity and selectivity of responses (such as CYP1A and ER α mRNA variations after OCs and PAHs exposure) and could represent a valid future approach for the study of inter- and intra-species sensitivities to various classes of environmental contaminants.
Product Description
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%