Merck
  • Capillary electrophoresis-mass spectrometry determination of morphine and its isobaric glucuronide metabolites.
Capillary electrophoresis-mass spectrometry determination of morphine and its isobaric glucuronide metabolites.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-16)
Theresa A Isbell, Erin C Strickland, Jennifer Hitchcock, Gregory McIntire, Christa L Colyer
ABSTRACT
The determination of morphine and its isobaric metabolites morphine-3-beta-d-glucuronide (M3G) and morphine-6-beta-d-glucuronide (M6G) is useful for therapeutic drug monitoring and forensic identification of drug use. In particular, capillary electrophoresis with mass spectrometry (CE-MS) represents an attractive tool for opioid analysis. Whereas volatile background electrolytes in CE often improve electrospray ionization for coupled MS detection, such electrolytes may reduce CE separation efficiency and resolution. To better understand the effects of background electrolyte (BGE) composition on separation efficiency and detection sensitivity, this work compares and contrasts method development for both volatile (ammonium formate and acetate) and nonvolatile (ammonium phosphate and borate) buffers. Peak efficiencies and migration times for morphine and morphine metabolites were optimal with a 25mM ammonium borate buffer (pH=9.5) although greater sensitivities were achieved in the ammonium formate buffer. Optimized CE methods allowed for the resolution of the isobaric morphine metabolites prior to high mass accuracy, electrospray ionization quadrupole time-of-flight (ESI-QTOF) MS detection applicable to the analysis of urine samples in under seven minutes. Urine sample preparation required only a 10-fold dilution with BGE prior to analysis. Limits of detection (LOD) in normal human urine were found to be 1.0μg/mL for morphine and 2.5μg/mL for each of M3G and M6G by CE-ESI-QTOF-MS. These LODs were comparable to those for CE-UV analysis of opioid standards in buffer, whereas CE-ESI-QTOF-MS analysis of opioid standards in buffer yielded LODs an order of magnitude lower. Patient urine samples (N=12) were analyzed by this new CE-ESI-QTOF-MS method and no significant difference in total morphine content relative to prior liquid chromatography-mass spectrometry (LC-MS) results was found as per a paired-t test at the 99% confidence level. Whereas the LC-MS method applied to these samples determined only total morphine content, this new CE-ESI-QTOF-MS method allowed for species differentiation in addition to total morphine determination. By this method, it was found that M3G and M6G metabolites were present in a 5:1 concentration ratio, on average, in patient samples. Therefore, the CE-ESI-QTOF-MS method not only allows for total morphine concentration determination comparable to established LC-MS methods, but also allows for differentiation between morphine and its trace glucuronides, yielding additional biochemical information about drug metabolism.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%