Merck
  • Cytosolic HMGB1 controls the cellular autophagy/apoptosis checkpoint during inflammation.
Cytosolic HMGB1 controls the cellular autophagy/apoptosis checkpoint during inflammation.
The Journal of clinical investigation (2015-02-03)
Xiaorong Zhu, Jeannette S Messer, Yunwei Wang, Fanfei Lin, Candace M Cham, Jonathan Chang, Timothy R Billiar, Michael T Lotze, David L Boone, Eugene B Chang
ABSTRACT
The intracellular protein HMGB1 is released from cells and acts as a damage-associated molecular pattern molecule during many diseases, including inflammatory bowel disease (IBD); however, the intracellular function of HMGB1 during inflammation is poorly understood. Here, we demonstrated that cytosolic HMGB1 regulates apoptosis by protecting the autophagy proteins beclin 1 and ATG5 from calpain-mediated cleavage during inflammation. Colitis in mice with an intestinal epithelial cell-specific Hmgb1 deletion and patients with IBD were both characterized by increased calpain activation, beclin 1 and ATG5 cleavage, and intestinal epithelial cell (IEC) death compared with controls. In vitro cleavage assays and studies of enteroids verified that HMGB1 protects beclin 1 and ATG5 from calpain-mediated cleavage events that generate proapoptotic protein fragments. Together, our results indicate that HMGB1 is essential for mitigating the extent and severity of inflammation-associated cellular injury by controlling the switch between the proautophagic and proapoptotic functions of beclin 1 and ATG5 during inflammation. Moreover, these studies demonstrate that HMGB1 is pivotal for reducing tissue injury in IBD and other complex inflammatory disorders.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Pricing and availability is not currently available.
SAFC
HEPES
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Pricing and availability is not currently available.
Sigma-Aldrich
Calpeptin, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Pricing and availability is not currently available.
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
SAFC
HEPES
Pricing and availability is not currently available.