Merck
  • Vitamin A metabolism and mucosal immune function are distinct between BALB/c and C57BL/6 mice.
Vitamin A metabolism and mucosal immune function are distinct between BALB/c and C57BL/6 mice.
European journal of immunology (2014-10-15)
Gera Goverse, Brenda J Olivier, Rosalie Molenaar, Marlene Knippenberg, Mascha Greuter, Tanja Konijn, Emma C L Cook, Marieke R Beijer, Dawn M Fedor, Joke M M den Haan, Joseph L Napoli, Gerd Bouma, Reina E Mebius
ABSTRACT
The vitamin A metabolite retinoic acid (RA) has been reported to suppress Th1 responses and enhance Th2 responses. Here, we investigated whether differences in vitamin A metabolism could underlie the differences between C57BL/6 and BALB/c mice, which are reportedly seen as Th1 and Th2 responders, respectively. BALB/c mice were shown to have higher intestinal epithelial expression of RALDH1 (where RALDH is retinaldehyde dehydrogenase), and, consequently, higher RALDH activity in MLN-DCs, leading to an increased ability to induce IgA class switching in B cells. Furthermore, within BALB/c mice, induction of IgA secretion as well as increased accumulation of regulatory T cells (Treg) in the intestinal lamina propria was observed. Additionally, as BALB/c mice are more resistant to dextran sulphate sodium (DSS) induced colitis, mice that lacked vitamin A in their diet had a more severe form of DSS-induced colitis compared to control mice. Therefore, the level of RA production and consequently the degree of RA-mediated signaling is crucial for the efficiency of the mucosal immune system.
Product Description
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%