Merck
Myrtus communis L. as source of a bioactive and safe essential oil.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-12-03)
Amel Bouzabata, Célia Cabral, Maria José Gonçalves, Maria Teresa Cruz, Ange Bighelli, Carlos Cavaleiro, Joseph Casanova, Félix Tomi, Ligia Salgueiro
ABSTRACT
In Algeria, Myrtus communis L. is distributed throughout the Tell Atlas and the coastal regions of Algiers and Constantine. The leaves are used in respiratory disorders, diarrhea and hemorrhoids. The aims of this work were to evaluate the antifungal and anti-inflammatory potential of well characterized essential oils (EO). Since EO can be applied by inhalation, dermal application and oral administration, we used several mammalian cell lines to assess safe bioactive doses. The chemical composition of two samples was investigated by GC-FID, GC-MS and (13)C NMR spectroscopy. Monoterpene derivatives are the main compounds: α-pinene (50.8 and 33.6%), 1,8-cineole (21.9 and 13.3%), linalool (2.7 and 14.8%), and linalyl acetate (0.5 and 9.5%). The antifungal evaluation revealed that the oils were more active against Cryptococcus neoformans (yeast) and Epidermophyton floccosum, Microsporum canis, Trichophyton rubrum (dermatophytes). The anti-inflammatory potential was evaluated using an in vitro model of lipopolysaccharide (LPS)-stimulated macrophages. Assessment of cell viability was made through the MTT assay. Both oils were able to significantly inhibit NO production, without affecting cell viability, in concentrations up to 0.64 mg/mL. These promising results, disclose bioactive concentrations of Myrtle essential oils with a safety profile suggesting a potential oral and topical application or use by inhalation.
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Supelco
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Sigma-Aldrich
Tetramethylsilane, NMR grade, ACS reagent, ≥99.9%
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard