Merck
  • Anti-IL-31 receptor antibody is shown to be a potential therapeutic option for treating itch and dermatitis in mice.
Anti-IL-31 receptor antibody is shown to be a potential therapeutic option for treating itch and dermatitis in mice.
British journal of pharmacology (2014-06-20)
K Kasutani, E Fujii, S Ohyama, H Adachi, M Hasegawa, H Kitamura, N Yamashita
ABSTRACT
IL-31, which is described as a pruritogenic cytokine, is linked to the itching that is associated with allergic and non-allergic eczema, but the precise pruritogenic mechanism of IL-31 and its potential as a therapeutic target for atopic dermatitis (AD) have not been determined. We investigated the effects of existing drugs on the scratching behaviour induced by an i.v. injection of IL-31 to clarify whether IL-31 induced pruritus indirectly. In addition, we studied the effects of an anti-IL-31 receptor α subunit (anti-IL-31 receptor α) neutralizing antibody on chronic pruritus-inducing dermatitis in an AD-like model to determine whether IL-31 not only induces scratching behaviour, but is also the causative factor in an AD phenotype. The scratching behaviour induced by an i.v. injection of IL-31 was inhibited by pretreatment with an anti-IL-31 receptor α-neutralizing antibody. In contrast, it was not inhibited significantly by a non-sedative antihistamine (terfenadine), immunosuppressants (dexamethasone and tacrolimus), or a μ-opioid receptor antagonist (naloxone). The anti-IL-31 receptor α-neutralizing antibody reduced the ear swelling and dermatitis score in a chronic pruritus-inducing AD-like model. Moreover, treatment with the anti-IL-31 receptor α-neutralizing antibody showed therapeutic effects on the dermatitis even if it was injected after the disease had developed. Anti-IL-31 receptor α is a potential novel therapeutic approach for escaping from the itch-scratch cycle and also a treatment for dermatitis in AD.
Product Description
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
FK-506 monohydrate, ≥98% (HPLC)
USP
Tacrolimus, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard