Merck
  • Reversible and irreversible small molecule inhibitors of monoamine oxidase B (MAO-B) investigated by biophysical techniques.
Reversible and irreversible small molecule inhibitors of monoamine oxidase B (MAO-B) investigated by biophysical techniques.
Bioorganic & medicinal chemistry (2015-01-21)
Rafael J Rojas, Dale E Edmondson, Terri Almos, Roderick Scott, Mark E Massari
ABSTRACT
Monoamine oxidase B (MAO-B) plays a key role in the metabolism of dopamine, a neurotransmitter critical for the maintenance of cognitive function. Consequently, MAO-B is an important therapeutic target for disorders characterized by a decline in dopaminergic neurotransmission, including Parkinson's disease (PD). An emerging strategy in drug discovery is to utilize the biophysical approaches of thermal shift and isothermal titration calorimetry (ITC) to gain insight into binding modality and identify thermodynamically privileged chemical scaffolds. Described here is the development of such approaches for reversible and irreversible small molecule inhibitors of MAO-B. Investigation of soluble recombinant MAO-B revealed mechanism-based differences in the thermal shift and binding thermodynamic profiles of MAO-B inhibitors. Irreversible inhibitors demonstrated biphasic protein melt curves, large enthalpically favorable and entropically unfavorable binding, in contrast to reversible compounds, which were characterized by a dose-dependent increase in thermal stability and enthalpically-driven binding. The biophysical approaches described here aim to facilitate the discovery of next-generation MAO-B inhibitors.
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Phenethylamine, ≥99%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
N-Benzylmethylamine, 97%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Phenethylamine, 99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material