Merck
  • Direct sample introduction gas chromatography and mass spectrometry for the determination of phthalate esters in cleaning products.
Direct sample introduction gas chromatography and mass spectrometry for the determination of phthalate esters in cleaning products.
Journal of chromatography. A (2015-01-15)
J I Cacho, N Campillo, P Viñas, M Hernández-Córdoba
ABSTRACT
A method using direct sample introduction (DSI) coupled to gas chromatography-mass spectrometry (GC-MS) is developed for the determination of six phthalate esters (dimethyl, diethyl, dibutyl, butylbenzyl, diethylhexyl and dioctyl phthalate) in cleaning products. The different variables involved in the DSI step, including venting time and temperature, vaporisation time and temperature, injector heating temperature and gas flow rate and pressure, were evaluated and optimised using Taguchi orthogonal arrays. The proposed method, using calibration against methanolic standards, showed good linearity in the 0.05-15 μg g(-1) range and good repeatability, with RSD values ranging from 3.5% to 5.7%. Quantification limits between 0.010 and 0.041 μg g(-1), depending on the compound, were attained, while recovery assays provided values from 83% to 115%. Twenty-seven cleaning products were analysed using the DSI-GC-MS method, being four phthalates (dimethyl, diethyl, dibutyl and diethylhexyl phthalate) found in fourteen of them at concentration levels in the 0.1-21 μg g(-1) range. Compared with the most common GC injection technique, which uses the split/splitless injector, the proposed DSI procedure provided larger peak areas and lower detection limits, as result of the greater injected volume and reduction in noise.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
Dimethyl phthalate, PESTANAL®, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Dess-Martin periodinane, 97%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%