Merck
  • MicroRNA let-7b suppresses human gastric cancer malignancy by targeting ING1.
MicroRNA let-7b suppresses human gastric cancer malignancy by targeting ING1.
Cancer gene therapy (2015-01-24)
X Han, Y Chen, N Yao, H Liu, Z Wang
ABSTRACT
MicroRNAs (miRNAs) are important regulators that play key roles in tumorigenesis and tumor progression. In this study, we investigate whether let-7b acts as a tumor suppressor to inhibit invasion and metastasis in gastric cancers. We analyzed the expression of let-7b in 60 pair-matched gastric neoplastic and adjacent non-neoplastic tissues by quantitative real-time polymerase chain reaction. Functional analysis of let-7b expression was assessed in vitro in gastric cancer cell lines with let-7b precursor and inhibitor. The roles of let-7b in tumorigenesis and tumor metastasis were analyzed using a stable let-7b expression plasmid in nude mice. A luciferase reporter assay was used to assess the effect of let-7b on inhibitor of growth family, member 1 (ING1) expression. Real-time PCR showed decreased levels of let-7b expression in metastatic gastric cancer tissues and cell lines that are potentially highly metastatic. Cell invasion and migration were significantly impaired in GC9811-P and SGC7901-M cell lines after transfection with let-7b mimics. Nude mice with xenograft models of gastric cancer confirmed that let-7b could inhibit gastric cancer metastasis in vivo after transfection by the lentivirus pGCsil-GFP- let-7b. Luciferase reporter assays demonstrated that let-7b directly binds to the 3'-UTR of ING1, and real-time PCR and western blotting further indicated that let-7b downregulated the expression of ING1 at the mRNA and protein levels. Our study demonstrates that overexpression of let-7b in gastric cancer can inhibit invasion and migration of gastric cancer cells through directly targeting the tumor metastasis-associated gene ING1. These findings help clarify the molecular mechanisms involved in gastric cancer metastasis and indicate that let-7b modulation may be a bona fide treatment of gastric cancer.
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%