Merck
  • In vitro antioxidant and anti-inflammatory activities of Radix Isatidis extract and bioaccessibility of six bioactive compounds after simulated gastro-intestinal digestion.
In vitro antioxidant and anti-inflammatory activities of Radix Isatidis extract and bioaccessibility of six bioactive compounds after simulated gastro-intestinal digestion.
Journal of ethnopharmacology (2014-09-27)
Ping Xiao, Hongzhi Huang, Jianwei Chen, Xiang Li
ABSTRACT
Radix Isatidis called "Ban-Lan-Gen" is one of the most commonly-used traditional Chinese medicines for antiviral, anti-inflammatory, antioxidant and antipyretic purposes. Investigate the bioaccessibility of uridine, epigoitrin, adenosine, clemastanin B, indigoticoside A and isolariciresinol as well as the antioxidant and anti-inflammatory activities during an in vitro gastro-intestinal digestion of the Radix Isatidis extract (RIE). High performance liquid chromatography (HPLC) technique was adopted to determine the bioaccessibility of six bioactive compounds in RIE. Antioxidant activities of RIE in different digestive stages were determined by 1,1-Diphenyl-2-picrylhydrazyl (DPPH), superoxide anion and hydroxyl radical scavenging abilities. Anti-inflammatory activity was assayed by the inhibitions of inflammatory cytokines such as nitrous oxide (NO), prostaglandin E2 (PGE2) and tumor necrosis factor α(TNF-α) producted by lipopolysaccharide (LPS) stimulated RAW264.7 cells. The bioaccessibility of uridine, epigoitrin, adenosine, clemastanin B, indigoticoside A and isolariciresinol were 15.38%, 18.28%, 24.01%, 6.50%, 8.65% and 17.78%, respectively. Also, the digestion products still possessed certain antioxidant activities. The antioxidant activity was highly correlated with lignans (clemastanin B, indigoticoside A and isolariciresino). The anti-inflammation activity of the three samples decreased in the order: IN sample (the solution that had diffused into the dialysis tubing)>Nondigested sample (RIE solution)>Gastric sample (post-gastric digestion)>OUT sample (material that remained in the gastro-intestinal tract). Results obtained in this research reveal the amount of bioactive compounds from RIE that could be available for absorption in vivo. The antioxidant activity decreased significantly but the anti-inflammatory activity was enhanced in serum-available fraction after gastro-intestinal digestion in vitro. This study could provide a scientific basis for a deeper pharmacological activity study of Radix Isatidis and a simple method for pharmacodynamic material basis research.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder