Merck
  • Effect of sample preparation on components and liver toxicity of Polygonum multiflorum.
Effect of sample preparation on components and liver toxicity of Polygonum multiflorum.
Journal of pharmaceutical and biomedical analysis (2015-03-15)
G P Lv, L Z Meng, D Q Han, H Y Li, J Zhao, S P Li
ABSTRACT
It was shown that different extracts had significant differences in the toxicity of Polygonum multiflorum. In this study, the effect of sample preparation on components and liver toxicity of different extracts from P. multiflorum were determined. Hepatoxic components were discovered based on biomembrane extraction. Comparative chemistry and toxicology between ethanol and water extracts were also performed. The results showed that ethanol extract had much stronger hepatotoxicity, the content of emodin-8-O-β-d-glucopyranoside, physcion-8-O-β-d-glucopyranoside, emodin and physcion was significantly higher in ethanol extract than in water extract, while the human hepatocytes extraction showed that 2,3,5,4'-tetrahydroxystilbene-2-O-β-d-glucopyranoside, emodin-8-O-β-d-glucopyranoside, physcion-8-O-β-d-glucopyranoside, emodin and physcion had interaction with human hepatocytes. The hepatotoxic effect of these components was investigated on human hepatocytes LO2 cells and emodin-8-O-β-d-glucopyranoside, physcion-8-O-β-d-glucopyranoside, emodin and physcion were finally confirmed to be, at least partial, hepatotoxic components. The results showed that sample preparation has significant effect on components in extracts of P. multiflorum especially the components related to hepatotoxicity. Water extract, the conventional administration form of Chinese herbs, is prefer for phytotherapy before well understanding their chemistry and biological activities.
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O