Merck
  • A quantitative ¹H nuclear magnetic resonance (qHNMR) method for assessing the purity of iridoids and secoiridoids.
A quantitative ¹H nuclear magnetic resonance (qHNMR) method for assessing the purity of iridoids and secoiridoids.
Fitoterapia (2014-12-17)
Zeyun Li, Edward Welbeck, Li Yang, Chunyong He, Haijun Hu, Ming Song, Kaishun Bi, Zhengtao Wang
ABSTRACT
This paper utilized a quantitative (1)H nuclear magnetic resonance (qHNMR) method for assessing the purity of iridoids and secoiridoids. The method was fully validated, including specificity, linearity, accuracy, precision, reproducibility, and robustness. For optimization of experimental conditions, several experimental parameters were investigated, including relaxation delay (D1), scan numbers (NS) and power length (PL1). The quantification was based on the area ratios of H-3 from analytes relative to aromatic protons from 1,4-dinitrobenzene (internal standard) with methanol-d4 as solvent. Five iridoids and secoiridoids (sweroside, swertiamarin, gentiopicroside, geniposide, genipin) were analyzed. Furthermore, the results were validated by the high performance liquid chromatography coupled with ultraviolet detection (HPLC-UV) method. It can be concluded that the qHNMR method was simple, rapid, and accurate, providing a reliable and superior method for assessing the purity of iridoids and secoiridoids.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, "100%", 99.96 atom % D
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, anhydrous, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, 99 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)